Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNAI3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568823
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568823 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568823 Supplier Invitrogen™ Supplier No. PA568823
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Human: 86%; Mouse: 100%; Rabbit: 85%; Rat: 100%.

SNAI3 is a member of the SNAIL gene family, named for the Drosophila snail gene, which plays roles in mesodermal formation during embryogenesis (Katoh and Katoh, 2003).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNAI3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SNAI3
Gene Accession No. Q3KNW1, Q9QY31
Gene Alias AI643946; Protein snail homolog 3; Smuc; SNAI3; snail family transcriptional repressor 3; snail family zinc finger 3; snail homolog 3; SNAIL3; snail-related gene from muscle cells; snail-related protein 2; Zfp293; zinc finger protein 293; Zinc finger protein SNAI3; ZNF293
Gene Symbols SNAI3
Host Species Rabbit
Immunogen synthetic peptide directed towards the n terminal region of mouse SNAI3.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30927, 333929
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.