Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Snail Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP229626FR
Description
Snail Polyclonal specifically detects Snail in Human, Mouse, Primate samples. It is validated for Western Blot.Specifications
Snail | |
Polyclonal | |
Western Blot | |
Protein sna, Protein snail homolog 1, SLUGH2, SNA, SNAHdJ710H13.1, snail 1 (drosophila homolog), zinc finger protein, snail 1 homolog, snail 1 zinc finger protein, snail 1, zinc finger protein, snail homolog 1 (Drosophila), zinc finger protein SNAI1 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Western Blot | |
DyLight 680 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SNAI1 | |
Synthetic peptide made to an internal portion of human SNAIL protein (between residues 200-300) [UniProt O95863] | |
0.1 mL | |
Breast Cancer, Cancer, Developmental Biology, Hypoxia, Lipid and Metabolism, Metastasis, Transcription Factors and Regulators | |
Human, Mouse, Primate | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction