Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ SnakeSkin™ Dialysis Tubing, 3.5K MWCO, 35 mm, 10.5 m PROMO

Catalog No. PI88244PM
missing translation for 'orderingAttributeHoverText'
:
Thermo Scientific™ SnakeSkin™ Dialysis Tubing

Perform desalting and buffer exchange for 10 to 100 mL samples with this 3.5K, 7K, and 10K molecular weight cut-off (MWCO), ready-to-use form of traditional dialysis membrane tubing. Securely close dialysis tubing up to 50 mm flat diameter with the sturdy, non-floating tube SnakeSkin Dialysis Clips.

The easy-open, pleated format of SnakeSkin Dialysis Tubing was developed to simplify large volume sample dialysis. To use, simply pull out and cut off the required length of tubing, fold over one end of tubing and apply a dialysis clip to close it, then add sample at the open end and use a second clip to close the remaining end. Clips are sold separately. In comparison, many traditional dialysis tubing membranes are supplied as dry, flattened rolls that are difficult to open and often require extensive soaking before they can be used.

The SnakeSkin membrane-pleating process does not change the tubing's MWCO. Also, any low MW contaminants present are removed during the dialysis process. SnakeSkin Dialysis Tubing is made from the same type of regenerated cellulose as flat tubing, so it matches the dialysis performance of conventional tubing.

Features of SnakeSkin Dialysis Tubing:

  • Convenient—ready to use, pre-wetted, pleated tube
  • High recovery—greater than 90% protein recovery
  • Speed—dialysis is generally completed in 4–6 hours
  • Stability—compatible with a variety of laboratory solutions, including acids, bases, hydrophobic solvents, and alcohols

Features of SnakeSkin Dialysis Tubing Clips:

  • Durable—polyamide composition for long-life
  • Dove-tail latch—prevent sample loss
  • Heat resistant—autoclave up to 134°C

SnakeSkin Dialysis Tubing, 3.5K or 10K MWCO, is composed of regenerated-cellulose and supplied as an open, pleated (telescoped) tube. It is supplied in 20-cm sticks containing 35 feet of dialysis tubing having a 16-, 22-, or 35-mm circular internal diameter (I.D.). With similar capacity to 10.5 meters of 23–50 mm diameter dry flat width tubing, the hydrated SnakeSkin Dialysis Tubing holds ∼2–10 mL of sample per centimeter of length.

SnakeSkin 7K MWCO dialysis tubing is supplied in 20-cm sticks containing 35 feet of dialysis tubing having a 22-mm circular internal diameter (I.D.). Equivalent in capacity to 10.5 meters of 34-mm diameter dry flat width tubing, the hydrated SnakeSkin Dialysis Tubing holds ∼3.7 mL of sample per centimeter of length.

3.5K MWCO tubing specifications:

  • Membrane type: regenerated cellulose
  • Tubing nominal dry thickness: 0.9 to 1.2 mil (22 to 30 μm)
  • Glycerol content: trace
  • Sulfur content: 0.1 to 0.15%
  • Heavy metals content: trace

7K MWCO tubing specifications:

  • Membrane type: regenerated cellulose
  • Tubing nominal dry thickness: 0.9 to 1.2 mil (22 to 30 μm)
  • Glycerol content: 13%
  • Sulfur content: 0.1 to 0.15%
  • Heavy metals content: trace

10K MWCO tubing specifications:

  • Membrane type: regenerated cellulose
  • Tubing nominal dry thickness: 0.9–1.2 mil (22–30 μm)
  • Glycerol content: 21%
  • Sulfur content: 0.1–0.15%
  • Heavy metals content: trace

Dialysis clip specifications:

  • Height: 10 mm
  • Width: 16 mm
  • Inner Length: 50 mm (maximum closure width)
  • Outer Length: 66 mm (end to end)
  • Composition: polyamide (nylon) derivative

Specifications

Product Type Dialysis Tubing
Product Line SnakeSkin™
MWCO 3.5 kDa
Membrane Cellulose Membrane, Dialysis
Material Cellulose
Format Tubing
Diameter (Metric) 35 mm I.D.
Purification Target Buffer Exchange, Protein
Sufficient For Approx. 48 mL per 5 cm length (plus 2.5 cm at each end for closure) yields 100 uses
Quantity 10.5 m
Content And Storage Upon receipt store at room temperature or 4°C. Keep in closed original packaging tube to prevent desiccation, which results in cracks or pinholes in the membrane. For best results, keep membrane hydrated by occasionally adding 1-2 drops of distilled water to the packaging tube.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.