Learn More
Invitrogen™ SNAP alpha Polyclonal Antibody

Description
Recommended positive controls: HepG2, mouse brain, rat brain. Predicted reactivity: Mouse (97%), Rat (97%), Xenopus laevis (89%), Pig (97%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | SNAP alpha |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | NAPA |
| Gene Accession No. | P54920, P54921, Q9DB05 |
| Gene Alias | 1500039N14Rik; alpha snap; alpha soluble NSF attachment protein; alpha-SNAP; Alpha-soluble NSF attachment protein; a-SNAP; AW209189; hydrocephaly with hop gait; hyh; Napa; N-ethylmaleimide sensitive fusion protein attachment protein alpha; N-ethylmaleimide-sensitive factor attachment protein alpha; N-ethylmaleimide-sensitive factor attachment protein, alpha; NSF attachment protein alpha; RA81; Snap; SNAPA; SNAP-alpha; SNARE |
| Gene Symbols | NAPA |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.