Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Safe-Lock Tubes 0.5 mL - Microtube
GSA_VAGREENER_CHOICE

Catalog No. E0030123603 Shop All Eppendorf North America Products
Change view
Click to view available options
Color:
Amber
Assorted
Blue
Clear
Green
Red
Yellow
Purity or Quality Grade:
Biopur™ (Sterile)
Eppendorf Quality
Forensic DNA Grade
PCR Clean (Non-sterile)
Packaging:
50 Tubes, Individually Packed
500 Tubes ( 5 Bags x 100 Tubes)
500 Tubes (1 Bag x 500 Tubes)
500 Tubes (10 Bags x 50 Tubes)
Max. RCF:
22,000 x g
30,000 x g
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Purity or Quality Grade Packaging Max. RCF
E0030123603 Clear Forensic DNA Grade 500 Tubes (10 Bags x 50 Tubes) 30,000 x g
05-402-93 Clear PCR Clean (Non-sterile) 500 Tubes (1 Bag x 500 Tubes) 30,000 x g
05-402-19 Amber PCR Clean (Non-sterile) 500 Tubes (1 Bag x 500 Tubes) 30,000 x g
05-402-18 Clear Eppendorf Quality 500 Tubes (1 Bag x 500 Tubes) 30,000 x g
05-402-20 Blue Eppendorf Quality 500 Tubes (1 Bag x 500 Tubes) 30,000 x g
05-402-21 Green Eppendorf Quality 500 Tubes (1 Bag x 500 Tubes) 30,000 x g
05-402-22 Red Eppendorf Quality 500 Tubes (1 Bag x 500 Tubes) 30,000 x g
05-402-23 Yellow Eppendorf Quality 500 Tubes (1 Bag x 500 Tubes) 30,000 x g
05-402-4 Assorted Eppendorf Quality 500 Tubes ( 5 Bags x 100 Tubes) 30,000 x g
05-402-24A Clear Biopur™ (Sterile) 50 Tubes, Individually Packed 22,000 x g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. E0030123603 Supplier Eppendorf™ Supplier No. 0030123603
  • PRODUCT PROMOTION
    $188.00
    $93.28 / Each of 1

    Save $94.72

    50% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Eppendorf™ Safe-Lock Tubes 0.5 mL - Microtube ensures secure sample storage. With a unique locking mechanism, these tubes prevent accidental opening, safeguarding precious samples. Ideal for PCR, these tubes offer reliability and ease in a compact design.

  • The hinged Eppendorf Safe-Lock lid prevents unintended opening during incubation and storage.
  • g-Safe™ technology, dependent on the Safe-Lock Tube variant (refer to the manual), ensures exceptional centrifugation stability up to 30,000 x g, preventing sample loss in the event of tube breakage and providing enhanced safety when handling hazardous samples.
  • Manufactured without the use of slip agents, plasticizers, and biocides, substances known to leach from plastic consumables into the sample and adversely impact bio-assay results.
  • Polypropylene construction offers high resistance to chemicals, mechanical stress, and extreme temperatures.
  • Precise lid sealing minimizes evaporation rates during long-term storage.
  • The large frosted lid and side surface facilitate easy labeling.
  • Ensured functionality in a temperature range from -86 °C to 100 °C.
  • Autoclavable when open (121°C, 20 min.) for effective sterilization.
  • Available in lot-certified PCR Clean, Eppendorf Forensic DNA Grade, and Biopur™ quality, ensuring tested purity.

Order Info

Assorted packages include 100 each of natural, blue, green, red and yellow

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Cap Style Hinged lid
Closure Color Colorless
Closure Material Polypropylene
Autoclavable Autoclavable
Capacity (Metric) 0.5 mL
Color Clear
Material Polypropylene
For Use With (Application) Incubation and storage
No. per Pack 500 Pk.
Packaging 500 Tubes (10 Bags x 50 Tubes)
Temperature Range (English) -122.8°F to +212°F
Temperature Range (Metric) -86°C to +100°C
Sterility Non-sterile
Purity or Quality Grade Forensic DNA Grade
Graduated Yes
Capacity (English) 0.02 oz.
Closure Type Safe-lock Lid
Max. RCF 30,000 x g
Shape Cylindrical
Product Type Locking Snap-cap Microtube
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.