Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SNAP Tag Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ CAB4255
Description
CAB4255 detects SNAP in samples. CAB4255 has been successfully used in Western Blot applications. The CAB4255 immunogen is purified recombinant SNAP.

Specifications
SNAP Tag | |
Polyclonal | |
Unconjugated | |
Rabbit | |
Protein A/G | |
RUO | |
Tag | |
Antibody | |
IgG |
Western Blot | |
1 mg/mL | |
BBS with no preservative; pH 8.4 | |
purified recombinant SNAP. | |
100 μg | |
Primary | |
-20°C | |
Liquid | |
Tag |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction