Learn More
SNAP-tag Rabbit, Polyclonal, GenScript™
Rabbit Polyclonal Antibody
Supplier: Genscript Corporation A0068440

Description
Reconstitute the lyophilized antibody with deionized water to the final concentration of 0.5 mg/mL. Well-characterized antibodies for epitope tags have been widely used in the study of protein expression in various systems. The SNAP-tag is a self-labeling protein tag that forms a highly stable, covalent thioether bond with fluorophores and other substituted groups on its benzyl guanine substrate. The expressed SNAP-tag fusion proteins can be used in many different assay formats depending upon the labeling substrate employed. GenScript Rabbit Anti-SNAP-tag Polyclonal Antibody is developed in rabbit hosts using purified recombinant SNAP-tag fusion protein. This polyclonal antibody is highly purified from rabbit antiserum by immunoaffinity chromatography.
SNAP-tag Polyclonal antibody specifically detects SNAP-tag in Tag samples. It is validated for ELISA, Western BlotSpecifications
SNAP-tag | |
Polyclonal | |
PBS with 0.02% sodium azide; pH 7.4 | |
Purified recombinant SNAP-tag fusion protein | |
40 μg | |
Primary | |
Antibody | |
IgG |
ELISA, Western Blot | |
Unconjugated | |
Rabbit | |
Affinity chromatography | |
RUO | |
-20°C or -80°C if preferred | |
Lyophilized | |
Tag |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.