Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNAP23 Antibody (2D8), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | SNAP23 |
---|---|
Clone | 2D8 |
Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
Applications | ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
SNAP23 Monoclonal antibody specifically detects SNAP23 in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
SNAP23 | |
ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
HsT17016, SNAP-23, SNAP23A, SNAP23B, synaptosomal-associated protein 23, synaptosomal-associated protein, 23kD, synaptosomal-associated protein, 23kDa, Vesicle-membrane fusion protein SNAP-23 | |
A synthesized peptide derived from Human SNAP23 [UniProt O00161] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
2D8 | |
ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neuroscience, Neurotransmission, Protein Phosphatase | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
8773 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title