Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNAP23 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA1738

Catalog No. PA1738


Only null left
Edge
Add to Cart

Description

Description

PA1-738 detects synaptosomal-associated protein 23 (SNAP-23) from human, mouse and rat samples. PA1-738 has been successfully used in Western blot and immunoprecipitation procedures. By Western blot, this antibody detects an ~23 kDa protein representing SNAP-23 from rat/mouse brain protein extract and HeLa cell lysates. PA1-738 immunizing peptide corresponds to amino acid residues 193-210 from mouse SNAP-23 protein. PA1-738 immunizing peptide (Cat. # PEP-147) is available for use in neutralization and control experiments.

Specificity of vesicular transport is regulated, in part, by the interaction of a vesicle-associated membrane protein termed synaptobrevin/VAMP with a target compartment membrane protein termed syntaxin. These proteins, together with SNAP25 (synaptosome-associated protein of 25 kDa), form a complex which serves as a binding site for the general membrane fusion machinery. Synaptobrevin/VAMP and syntaxin are believed to be involved in vesicular transport in most, if not all cells, while SNAP25 is present almost exclusively in the brain, suggesting that a ubiquitously expressed homolog of SNAP25 exists to facilitate transport vesicle/target membrane fusion in other tissues. The protein encoded by this gene is structurally and functionally similar to SNAP25 and binds tightly to multiple syntaxins and synaptobrevins/VAMPs. It is an essential component of the high affinity receptor for the general membrane fusion machinery and is an important regulator of transport vesicle docking and fusion. Two alternative transcript variants encoding different protein isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SNAP23
Polyclonal
Unconjugated
SNAP23
23kDa; AA408749; HsT17016; OTTHUMP00000161263; SNAP23; SNAP-23; SNAP23A; SNAP23B; Sndt; Synaptosomal-associated protein 23; synaptosomal-associated protein 23 kD; synaptosomal-associated protein, 23 kD; synaptosomal-associated protein, 23kD; synaptosomal-associated protein, 23kDa; synaptosome associated protein 23; synaptosome associated protein 23kDa; Syndet; Vesicle-membrane fusion protein SNAP-23
Rabbit
Antigen Affinity Chromatography
RUO
20619, 64630, 8773
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunoprecipitation, Western Blot, Immunocytochemistry
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
O00161, O09044, O70377
SNAP23
Sythetic Peptide: C N(193) K N R I D I A N T R A K K L I D S(210).
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.