Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNAP23 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501726630 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-6630 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-6630 Supplier Proteintech Group Inc Supplier No. 108251AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Synaptosomal-associated proteins (SNAPs) are cytosolic proteins that play a key role in in the process of membrane fusion in intracellular vesicle trafficking. In eukaryotic cells, the SNARE (soluble N-ethylmaleimide-sensitive factor attachment protein receptor) complex is critical to membrane docking and fusion and is believed to impart some degree of specificity between vesicle SNARE (v-SNARE) and target organelle SNARE (t-SNARE). In neurons and neuroendocrine cells, the SNARE complex consists of the integral membrane proteins VAMP (vesicle-associated membrane protein), syntaxin and SNAP-25. In non-neuronal tissue, a SNAP-25 homolog, SNAP-23, functionally replaces SNAP-25 in the SNARE complex. Studies show that VAMP, syntaxin and SNAP-23 are required for SNARE function and that this complex exists as a heterotrimer of the three proteins. In insulin signaling pathways, studies suggest that the translocation of GLUT4 in adipocytes requires that functional SNAP-23 be present in SNARE complexes.

Specifications

Antigen SNAP23
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Concentration 0.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene SNAP23
Gene Accession No. O00161, O09044, O70377
Gene Alias HsT17016, SNAP 23, SNAP23, SNAP23A, SNAP23B
Gene Symbols Snap23
Host Species Rabbit
Immunogen SNAP23 Fusion Protein Ag1269
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20619, 64630, 8773
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.