Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNAP25 Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NB1001492
Description
SNAP25 Polyclonal specifically detects SNAP25 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
SNAP25 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
bA416N4.2, dJ1068F16.2, FLJ23079, resistance to inhibitors of cholinesterase 4 homolog, RIC4, RIC-4, SEC9, SNAP-25SNAP, SUP, Super protein, Synaptosomal-associated 25 kDa protein, synaptosomal-associated protein 25, synaptosomal-associated protein, 25kD, synaptosomal-associated protein, 25kDa | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is expected to recognize both reported isoforms NP_003072.2 (SNAP25A) and NP_570824.1 (SNAP25B). Reported variants represent identical protein: NP_001309831.1, NP_003072.2 Reported variants represent identical protein: NP_001309834.1, NP_001309832.1, NP_001309833.1, NP_001309839.1, NP_001309836.1, NP_570824.1, NP_001309838.1, NP_001309835.1, NP_001309837.1 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
0.5 mg/mL | |
Western Blot 0.03-0.1 ug/ml, Immunohistochemistry 2 - 4 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500, Peptide ELISA detection limit 1:32000 | |
NP_003072.2 | |
SNAP25 | |
Peptide with sequence C-DEANQRATKMLGSG corresponding to C-Terminus according to NP_003072.2, NP_570824.1. | |
0.1 mg | |
Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision | |
6616 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction