Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNAP25 Monoclonal Antibody (OTI4C6), TrueMAB™, OriGene
SDP

Catalog No. 502541197
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1197 30 μL
50-167-3958 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1197 Supplier OriGene Supplier No. TA502964S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SNAP25 Monoclonal antibody specifically detects SNAP25 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen SNAP25
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI4C6
Concentration 0.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Snap25
Gene Accession No. P60880
Gene Alias ALMS1; bA416N4.2; Bdr; blind drunk; CG17676; CG17884; CG40452; CMS18; dJ1068F16.2; Dmel CG40452; Dmel_CG40452; etID17571.20; etID69167.20; fj42b02; fj45g04; fj48c02; FLJ23079; GENA70; HGNC:11132; resistance to inhibitors of cholinesterase 4 homolog; RIC4; RIC-4; SEC9; si:dkeyp-8f4.6; Snap; SNAP 25; snap25; SNAP-25; snap25.1; SNAP-25.1; snap25a; SNAP-25A; SNAP-25B; snp25; sp; SUP; super protein; synaptosomal associated protein-25; synaptosomal-associated 25 kDa protein; Synaptosomal-associated protein 25; synaptosomal-associated protein 25-A; synaptosomal-associated protein, 25 kDa; synaptosomal-associated protein, 25a; synaptosomal-associated protein, 25kDa; synaptosome associated protein 25; synaptosome associated protein 25a; synaptosome associated protein 25kDa; synaptosome-associated protein 25.1; synaptosome-associated protein 25a; wu:fj42b02; wu:fj45g04; wu:fj48c02
Host Species Mouse
Immunogen Full length human recombinant protein of human SNAP25 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6616
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.