Learn More
SNAP25 Mouse anti-Human, Clone: OTI4C6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF502964

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
SNAP25 is a t-SNARE involved in the molecular regulation of neurotransmitter release. It may play a role in the synaptic function of specific neuronal systems. SNAP25 associates with proteins involved in vesicle docking and membrane fusion. It also regulates plasma membrane recycling through its interaction with CENPF. Mutations in the gene can results in myasthenic syndrome.Specifications
SNAP25 | |
Monoclonal | |
Unconjugated | |
SNAP25 | |
CMS18, RIC-4, RIC4, SEC9, SNAP, SNAP-25, SUP, bA416N4.2, dJ1068F16.2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
6616 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI4C6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P60880 | |
SNAP25 | |
Full length human recombit protein of human SNAP25 produced in HEK293 cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.