Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNAP29 Antibody (SR1445), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | SNAP29 |
---|---|
Clone | SR1445 |
Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1:50 |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Description
SNAP29 Monoclonal antibody specifically detects SNAP29 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
SNAP29 | |
Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
CEDNIKSoluble 29 kDa NSF attachment protein, SNAP-29FLJ21051, synaptosomal-associated protein 29, synaptosomal-associated protein, 29kD, synaptosomal-associated protein, 29kDa, Vesicle-membrane fusion protein SNAP-29 | |
A synthesized peptide derived from human SNAP29 (Uniprot #: O95721) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1445 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Neuronal Cell Markers, Neurotransmission | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
9342 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title