Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNAP47 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555782
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555782 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555782 Supplier Invitrogen™ Supplier No. PA555782
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DDIKVHSPYE ISIRQRFIGK PDMAYRLISA KMPEVIPILE VQFSKKMELL EDALVLRSAR TSSPAEKSCS VWHAASGLMG RTLHREPPAG DQE Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 81%.

SNAP47 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNAP47
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Snap47
Gene Accession No. Q5SQN1
Gene Alias 1110031B06Rik; C1orf142; C7H1orf142; CH1073-162B20.3; epididymis luminal protein 170; epididymis secretory protein Li 290; ESFI5812; HEL170; HEL-S-290; LOW QUALITY PROTEIN: synaptosomal-associated protein 47; MGC72593; RGD735194; snap47; SNAP-47; SVAP1; Synaptosomal-associated 47 kDa protein; synaptosomal-associated protein 47; synaptosomal-associated protein, 47; synaptosomal-associated protein, 47kDa; synaptosome associated protein 47; synaptosome associated protein 47kDa; Unknown (protein for MGC:153222); zgc:153222
Gene Symbols Snap47
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SNAP47. Recombinant protein control fragment (Product #RP-94298).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116841
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.