Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNAP47 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | SNAP47 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
SNAP47 Polyclonal specifically detects SNAP47 in Human samples. It is validated for Western Blot.Specifications
| SNAP47 | |
| Polyclonal | |
| Rabbit | |
| Q5SQN1 | |
| 116841 | |
| Synthetic peptides corresponding to C1ORF142 The peptide sequence was selected from the middle region of C1ORF142 (NP_444280). Peptide sequence TALHLQTSLPALSEADTQELTQILRRMKGLALEAESELERQDEALDGVAA. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| C1orf142, Epididymis luminal protein 170, FLJ12517, HEL170, SNAP-47chromosome 1 open reading frame 142, SVAP1DKFZp686M10160, Synaptosomal-associated 47 kDa protein, synaptosomal-associated protein 47, synaptosomal-associated protein, 47kDa | |
| SNAP47 | |
| IgG | |
| 52.5 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title