Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNAP91 Recombinant Rabbit Monoclonal Antibody (HL1657)
GREENER_CHOICE

Catalog No. PIMA547127
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547127 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547127 Supplier Invitrogen™ Supplier No. MA547127
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

Clathrin coat assembly protein AP180 is a protein that in humans is encoded by the SNAP91 gene. Adaptins are components of the adapter complexes which link clathrin to receptors in coated vesicles. Clathrin-associated protein complexes are believed to interact with the cytoplasmic tails of membrane proteins, leading to their selection and concentration. Binding of AP180 to clathrin triskelia induces their assembly into 60-70 nm coats.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNAP91
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone HL1657
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Snap91
Gene Accession No. O60641, Q05140, Q61548
Gene Alias 91 kDa synaptosomal-associated protein; 91kDa; Ap180; assembly protein 180 (AP180); assembly protein, 180kDa; CALM; Clathrin coat assembly protein AP180; clathrin coat-associated protein AP180; F1-20; KIAA0656; mKIAA0656; phosphoprotein F1-20; SNAP91; synaptosomal-associated protein 91; synaptosomal-associated protein 91 kDa; synaptosomal-associated protein, 91 kDa; synaptosomal-associated protein, 91kDa homolog; synaptosome associated protein 91; synaptosome associated protein 91kDa
Gene Symbols Snap91
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of mouse SNAP91. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20616, 65178, 9892
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.