Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNAP91 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556217
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556217 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556217 Supplier Invitrogen™ Supplier No. PA556217
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: APSEGSAEAA PELDLFAMKP PETSVPVVTP TASTAPPVPA TAPSPAPAVA AAAAATTAAT AAATTTTTTS AATATTAPPA LDIFGDLFES TP Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.

Clathrin coat assembly protein AP180 is a protein that in humans is encoded by the SNAP91 gene. Adaptins are components of the adapter complexes which link clathrin to receptors in coated vesicles. Clathrin-associated protein complexes are believed to interact with the cytoplasmic tails of membrane proteins, leading to their selection and concentration. Binding of AP180 to clathrin triskelia induces their assembly into 60-70 nm coats.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNAP91
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Snap91
Gene Accession No. O60641
Gene Alias 91 kDa synaptosomal-associated protein; 91kDa; Ap180; assembly protein 180 (AP180); assembly protein, 180kDa; CALM; Clathrin coat assembly protein AP180; clathrin coat-associated protein AP180; F1-20; KIAA0656; mKIAA0656; phosphoprotein F1-20; SNAP91; synaptosomal-associated protein 91; synaptosomal-associated protein 91 kDa; synaptosomal-associated protein, 91 kDa; synaptosomal-associated protein, 91kDa homolog; synaptosome associated protein 91; synaptosome associated protein 91kDa
Gene Symbols Snap91
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SNAP91. Recombinant protein control fragment (Product #RP-94629).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9892
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.