Learn More
Invitrogen™ SNAP91 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA593191
Description
Immunogen sequence: AVPVSTSKPS SDLLDLQPDF SSGGAAAAAA PAPPPPAGGA TAWGGFGGSF MAPSPSPVTP AQNNLLQPNF EAAFGTTPST SSSSSFDPSV FDGLGDLLMP TMAPAGQPAP VSMVPPSPAM AASKALGSDL DSSLASLVGN LGISGTTTKK GDLQWNAGEK KLTGGANWQP KVAPATWSAG VPPSAPLQGA VPPTSSVPPV AGAPSVGQPG AGFGMPPAGT GMPMMPQQPV MFAQPMMRPP FGAAAVPGTQ LSPSPTPASQ SPKKPPAKDP LADLNIKDFL; Positive Samples: Mouse brain, Rat brain, Rat liver; Cellular Location: Cell membrane, Cytoplasmic side, Membrane, Peripheral membrane protein, coated pit.
Clathrin coat assembly protein AP180 is a protein that in humans is encoded by the SNAP91 gene. Adaptins are components of the adapter complexes which link clathrin to receptors in coated vesicles. Clathrin-associated protein complexes are believed to interact with the cytoplasmic tails of membrane proteins, leading to their selection and concentration. Binding of AP180 to clathrin triskelia induces their assembly into 60-70 nm coats.
Specifications
SNAP91 | |
Polyclonal | |
Unconjugated | |
Snap91 | |
91 kDa synaptosomal-associated protein; 91kDa; Ap180; assembly protein 180 (AP180); assembly protein, 180kDa; CALM; Clathrin coat assembly protein AP180; clathrin coat-associated protein AP180; F1-20; KIAA0656; mKIAA0656; phosphoprotein F1-20; SNAP91; synaptosomal-associated protein 91; synaptosomal-associated protein 91 kDa; synaptosomal-associated protein, 91 kDa; synaptosomal-associated protein, 91kDa homolog; synaptosome associated protein 91; synaptosome associated protein 91kDa | |
Rabbit | |
Affinity chromatography | |
RUO | |
20616, 65178 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q05140, Q61548 | |
Snap91 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 321-600 of human SNAP91 (NP_0012297231). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.