Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNAPC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584892
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584892 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584892 Supplier Invitrogen™ Supplier No. PA584892
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TPPGLQTDCE ALLSRFQETD SVRFEDFTEL WRNMKFGTIF CGRMRNLEKN MFTKEALALA WR Highest antigen sequence indentity to the following orthologs: Mouse - 87%, Rat - 84%.

SNAPC1 is a part of the SNAPc complex required for the transcription of both RNA polymerase II and III small-nuclear RNA genes. It binds to the proximal sequence element (PSE), a non-TATA-box basal promoter element common to these 2 types of genes and recruits TBP and BRF2 to the U6 snRNA TATA box.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNAPC1
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SNAPC1
Gene Accession No. Q16533
Gene Alias 2700033G17Rik; AU015787; Proximal sequence element-binding transcription factor subunit gamma; PSE-binding factor subunit gamma; PTF subunit gamma; PTFgamma; small nuclear RNA activating complex polypeptide 1; small nuclear RNA activating complex, polypeptide 1; small nuclear RNA activating complex, polypeptide 1, 43kD; small nuclear RNA activating complex, polypeptide 1, 43kDa; small nuclear RNA-activating complex polypeptide 1; SNAP43; SNAPc 43 kDa subunit; SNAPc subunit 1; SNAPC1; snRNA-activating protein complex 43 kDa subunit; snRNA-activating protein complex subunit 1
Gene Symbols SNAPC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SNAPC1. Recombinant protein control fragment (Product #RP-108597).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6617
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.