Learn More
Invitrogen™ SNAPC1 Polyclonal Antibody

Description
Peptide sequence: KMSLRNKGNV QNIHKEDKPL SLSMPVITEE EENESLSGTE FTASKKRRKH Sequence homology: Human: 100%.
Specifications
Specifications
| Antigen | SNAPC1 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | SNAPC1 |
| Gene Accession No. | Q16533 |
| Gene Alias | 2700033G17Rik; AU015787; Proximal sequence element-binding transcription factor subunit gamma; PSE-binding factor subunit gamma; PTF subunit gamma; PTFgamma; small nuclear RNA activating complex polypeptide 1; small nuclear RNA activating complex, polypeptide 1; small nuclear RNA activating complex, polypeptide 1, 43kD; small nuclear RNA activating complex, polypeptide 1, 43kDa; small nuclear RNA-activating complex polypeptide 1; SNAP43; SNAPc 43 kDa subunit; SNAPc subunit 1; SNAPC1; snRNA-activating protein complex 43 kDa subunit; snRNA-activating protein complex subunit 1 |
| Gene Symbols | SNAPC1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.