Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNAPC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565585
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565585 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565585 Supplier Invitrogen™ Supplier No. PA565585
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ALVEHMTETYL RLTAPQPIPA GGSLGPAAEG DGAGSKAPEE TPPATEKAEH SELKSPWQAA GICPLNPFLV PLELLGRA Highest antigen sequence identity to the following orthologs - mouse 51%, rat 49%.

This gene encodes a subunit of the snRNA-activating protein complex which is associated with the TATA box-binding protein. The encoded protein is necessary for RNA polymerase II and III dependent small-nuclear RNA gene transcription. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNAPC2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SNAPC2
Gene Accession No. Q13487
Gene Alias 0610007H10Rik; AU015675; Proximal sequence element-binding transcription factor subunit delta; PSE-binding factor subunit delta; PTF subunit delta; PTFDELTA; small nuclear RNA activating complex polypeptide 2; small nuclear RNA activating complex, polypeptide 2; small nuclear RNA activating complex, polypeptide 2, 45kD; small nuclear RNA activating complex, polypeptide 2, 45kDa; small nuclear RNA-activating complex polypeptide 2; SNAP45; SNAPc 45 kDa subunit; SNAPc subunit 2; SNAPC2; snRNA-activating protein complex 45 kDa subunit; snRNA-activating protein complex subunit 2
Gene Symbols SNAPC2
Host Species Rabbit
Immunogen Recombinant Human SNAPC2. Recombinant protein control fragment (Product #RP-106249).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6618
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.