Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNAPC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571460
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571460 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571460 Supplier Invitrogen™ Supplier No. PA571460
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Human: 100%.

This gene encodes a subunit of the snRNA-activating protein complex which is associated with the TATA box-binding protein. The encoded protein is necessary for RNA polymerase II and III dependent small-nuclear RNA gene transcription. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNAPC2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SNAPC2
Gene Accession No. Q13487
Gene Alias 0610007H10Rik; AU015675; Proximal sequence element-binding transcription factor subunit delta; PSE-binding factor subunit delta; PTF subunit delta; PTFDELTA; small nuclear RNA activating complex polypeptide 2; small nuclear RNA activating complex, polypeptide 2; small nuclear RNA activating complex, polypeptide 2, 45kD; small nuclear RNA activating complex, polypeptide 2, 45kDa; small nuclear RNA-activating complex polypeptide 2; SNAP45; SNAPc 45 kDa subunit; SNAPc subunit 2; SNAPC2; snRNA-activating protein complex 45 kDa subunit; snRNA-activating protein complex subunit 2
Gene Symbols SNAPC2
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human SNAPC2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6618
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.