Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNAPC3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988570
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8570 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8570 Supplier Bioss Supplier No. BS8452R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SNAPC3 is part of the SNAPc complex required for the transcription of both RNA polymerase II and III small-nuclear RNA genes. SNAPC3 binds to the proximal sequence element (PSE), a non-TATA-box basal promoter element common to these 2 types of genes. SNAPC3 recruits TBP and BRF2 to the U6 snRNA TATA box.

Specifications

Antigen SNAPC3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SNAPC3
Gene Accession No. Q92966
Gene Alias 1810020H02Rik; 4930558A07Rik; 5031401C21Rik; AI414457; BOS_8759; E030018J20Rik; Proximal sequence element-binding transcription factor subunit beta; PSE-binding factor subunit beta; PTF subunit beta; PTFbeta; small nuclear RNA activating complex polypeptide 3; small nuclear RNA activating complex, polypeptide 3; small nuclear RNA activating complex, polypeptide 3, 50kD; small nuclear RNA activating complex, polypeptide 3, 50kDa; Small nuclear RNA-activating complex polypeptide 3; SNAP50; SNAPc 50 kDa subunit; SNAPc subunit 3; Snapc3; snRNA-activating protein complex 50 kDa subunit; snRNA-activating protein complex subunit 3
Gene Symbols SNAPC3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SNAPC3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6619
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.