Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNAPC4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | SNAPC4 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
SNAPC4 Polyclonal specifically detects SNAPC4 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
SNAPC4 | |
Polyclonal | |
Rabbit | |
Human | |
6621 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MDVDAEREKITQEIKELERILDPGSSGSHVEISESSLESDSEADSLPSEDLDPADPPISEEERWGEASNDEDDPKDKTLPEDPETCLQLNMVYQEVIQE | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
FLJ13451, Proximal sequence element-binding transcription factor subunit alpha, PSE-binding factor subunit alpha, PTF subunit alpha, PTFalpha, small nuclear RNA activating complex, polypeptide 4, 190kD, small nuclear RNA activating complex, polypeptide 4, 190kDa, SNAP190SNAPc 190 kDa subunit, SNAPc subunit 4, snRNA-activating protein complex 190 kDa subunit, snRNA-activating protein complex subunit 4 | |
SNAPC4 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title