Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SNAPC4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100760
Description
Antibody detects endogenous levels of total SNAPC4.
Part of the SNAPc complex required for the transcription of both RNA polymerase II and III small-nuclear RNA genes. Binds to the proximal sequence element (PSE), a non-TATA-box basal promoter element common to these 2 types of genes. Recruits TBP and BRF2 to the U6 snRNA TATA box.
Specifications
SNAPC4 | |
Polyclonal | |
Unconjugated | |
SNAPC4 | |
GTAR; KIAA0697; proximal sequence element-binding transcription factor subunit alpha; PSE-binding factor subunit alpha; PTF subunit alpha; PTFalpha; small nuclear RNA activating complex polypeptide 4; small nuclear RNA activating complex, polypeptide 4; small nuclear RNA activating complex, polypeptide 4, 190kD; small nuclear RNA activating complex, polypeptide 4, 190kDa; SNAP190; SNAPc 190 kDa subunit; SNAPc subunit 4; SNAPC4; snRNA-activating protein complex 190 kDa subunit; snRNA-activating protein complex subunit 4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
362088, 6621 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q5SXM2 | |
SNAPC4 | |
A synthesized peptide derived from human SNAPC4(Accession Q5SXM2), corresponding to amino acid residues P53-E103. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction