Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNAPC5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103285
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103285 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103285 Supplier Invitrogen™ Supplier No. PA5103285
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SNAPC5.

Part of the SNAPc complex required for the transcription of both RNA polymerase II and III small-nuclear RNA genes. Binds to the proximal sequence element (PSE), a non-TATA-box basal promoter element common to these 2 types of genes. Recruits TBP and BRF2 to the U6 snRNA TATA box.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNAPC5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SNAPC5
Gene Accession No. O75971, Q8R2K7
Gene Alias 2010103A03Rik; CTD-3185P2.1; small nuclear RNA activating complex polypeptide 5; small nuclear RNA activating complex, polypeptide 5; small nuclear RNA activating complex, polypeptide 5, 19kD; small nuclear RNA activating complex, polypeptide 5, 19kDa; small nuclear RNA-activating complex polypeptide 5; SNAP19; SNAPc 19 kDa subunit; SNAPc subunit 5; SNAPC5; snRNA-activating protein complex 19 kDa subunit; snRNA-activating protein complex subunit 5
Gene Symbols SNAPC5
Host Species Rabbit
Immunogen A synthesized peptide derived from human SNAPC5(Accession O75971), corresponding to amino acid residues L22-N72.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10302, 330959
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.