Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SNAPC5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555068
Description
Immunogen sequence: MLSRLQELRK EEETLLRLKA ALHDQLNRLK VEELALQSMI SSRRGDEMLS SHTVPEQSHD MLVHVDNEAS INQTTLELST KSHVTEEEEE EEEEESDS Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 79%.
Part of the SNAPc complex required for the transcription of both RNA polymerase II and III small-nuclear RNA genes. Binds to the proximal sequence element (PSE), a non-TATA-box basal promoter element common to these 2 types of genes. Recruits TBP and BRF2 to the U6 snRNA TATA box.
Specifications
| SNAPC5 | |
| Polyclonal | |
| Unconjugated | |
| SNAPC5 | |
| 2010103A03Rik; CTD-3185P2.1; small nuclear RNA activating complex polypeptide 5; small nuclear RNA activating complex, polypeptide 5; small nuclear RNA activating complex, polypeptide 5, 19kD; small nuclear RNA activating complex, polypeptide 5, 19kDa; small nuclear RNA-activating complex polypeptide 5; SNAP19; SNAPc 19 kDa subunit; SNAPc subunit 5; SNAPC5; snRNA-activating protein complex 19 kDa subunit; snRNA-activating protein complex subunit 5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10302 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.8 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75971 | |
| SNAPC5 | |
| Recombinant protein corresponding to Human SNAPC5. Recombinant protein control fragment (Product #RP-89424). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction