Learn More
Applied Biosystems™ SNaPshot™ Multiplex Kit
Description
Includes
SNaPshot™ multiplex ready reaction mix (for 100 rxns), SNaPshot™ multiplex control primer mix (30μL), SNaPshot™ multiplex control template (60μL), protocol
- Perform multiplexed, single-base extension of up to 10 SNPs
- Interrogate multiple SNPs regardless of chromosome position
- Separate SNP loci that differ by a single base pair
- Perform robust interrogation of multiple SNPs with only a small amount of amplified template
- Decrease expenses with this moderately priced system
- Reduce the complexity of your experiments with this well-tested, easy-to-understand single-base extension technique
Order Info
Each
Specifications
Specifications
| Content And Storage | Contains: • SNaPshot™ Multiplex Ready Reaction Mix (for 100 reactions) • SNaPshot™ Multiplex Control Primer Mix (30 μl) • SNaPshot™ Multiplex Control Template (60 μl) • Protocol Store at -15°C to -25°C in a constant-temperature freezer. |
| Detection Method | Primer-probe |
| Format | Kit |
| Reaction Speed | 9 hr. |
| Technique | SNP Genotyping (Fragment Analysis-Based), Primer Extension |
| For Use With (Equipment) | 3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers, SeqStudio Genetic Analyzer, 3730/3730xl Genetic Analyzers |
| Product Line | SNaPshot |
| Product Type | Multiplex Kit |
| Quantity | 100 reactions |
| Run Time | 9 hr. |
| Show More |
Frequently Asked Questions (FAQs)
The SNaPshot Multiplex Kis is designed to interrogate up to 10 single nucleotide polymorphisms (SNPs) at known locations on 1-10 templates in a single tube.
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.