Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNAT1 Neurotransmitter Transporter Monoclonal Antibody (S104-32), QED Bioscience™
SDP

Catalog No. 502278217
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-8217 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-8217 Supplier Leinco Technologies Inc Supplier No. 56522100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

SNAT1 Neurotransmitter Transporter Monoclonal antibody specifically detects SNAT1 Neurotransmitter Transporter in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blot

Specifications

Antigen SNAT1 Neurotransmitter Transporter
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone S104-32
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Gene SLC38A1
Gene Accession No. Q8K2P7, Q9H2H9, Q9JM15
Gene Alias AA408026; AA409865; AL022800; Amino acid transporter A1; amino acid transporter system A1; ATA1; AU015942; GlnT; glutamine transporter; MNat2; Nat2; N-system amino acid transporter 2; rATA1; S38A1; Sa2; SAT1; SLC38A1; SNAT 1; Snat1; sodium-coupled neutral amino acid transporter 1; Solute carrier family 38 member 1; solute carrier family 38, member 1; system A amino acid transporter 1; System A transporter 2; system N amino acid transporter 1
Host Species Mouse
Immunogen Fusion protein corresponding to aa 1-63 (cytoplasmic N-terminus) of rat SNAT1 (accession no. NP_620187.1).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 105727, 170567, 81539
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.