Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNAT2 Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABN1734
Description
Anti-SNAT2 is a highly specific rabbit polyclonal antibody that targets Sodium-coupled neutral amino acid transporter 2 and has been tested in Immunocytochemistry, Immunofluorescence, Immunohistochemistry, and Western Blotting.
Specifications
SNAT2 | |
Polyclonal | |
Unconjugated | |
Rabbit | |
Unpurified | |
RUO | |
Primary | |
This polyclonal antibody detects sodium-coupled neutral amino acid transporter 2 (SNAT2) in rat brain. It targets an epitope within the first 65 amino acids. | |
Stable for 1 year at -20°C from date of receipt. |
Immunohistochemistry, Western Blot, Immunocytochemistry, Immunofluorescence | |
Please refer to lot specific datasheet. | |
Slc38a2,Ata2,Sa1,Sat2,Snat2 | |
GST-tagged recombinant fragment corresponding to the first 65 amino acids from te N-terminal region of rat Sodium-coupled neutral amino acid transporter 2 (SNAT2). | |
100 μL | |
Neuroscience | |
NP_851604 | |
Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction