Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNAT2 Rabbit anti-Rat, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABN173425UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN1734-25UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1734-25UL Supplier MilliporeSigma Supplier No. ABN173425UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-SNAT2, Cat. No. ABN1734, is a highly specific rabbit polyclonal antibody that targets Sodium-coupled neutral amino acid transporter 2 and has been tested in Immunocytochemistry, Immunofluorescence, Immunohistochemistry, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNAT2
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Rabbit polyclonal antiserum with 0.05% sodium azide.
Gene Alias Sodium-coupled neutral amino acid transporter 2;Amino acid transporter A2;Solute carrier family 38 member 2;System A amino acid transporter 2;System A transporter 1;System N amino acid transporter 2
Gene Symbols Slc38a2;Ata2;Sa1;Sat2;Snat2
Host Species Rabbit
Immunogen GST-tagged recombinant fragment corresponding to the first 65 amino acids from te N-terminal region of rat Sodium-coupled neutral amino acid transporter 2 (SNAT2).
Molecular Weight of Antigen 55.55 kDa calculated.
Purification Method Unpurified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity This polyclonal antibody detects sodium-coupled neutral amino acid transporter 2 (SNAT2) in rat brain. It targets an epitope within the first 65 amino acids.
Target Species Rat
Content And Storage Stable for 1 year at -20°C from date of receipt.
Product Type Antibodies
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.