Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SND1-IT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570068
Description
SND1-IT1 Polyclonal Antibody for Western Blot
SND1-IT1 is a protein coding gene.
Specifications
SND1-IT1 | |
Polyclonal | |
Unconjugated | |
SND1-IT1 | |
C7orf54; NAG8; NSG-X; SND1 intronic transcript 1; SND1-IT1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
27099 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9HBX3 | |
SND1-IT1 | |
synthetic peptide directed towards the N-terminal region of human SND1-IT1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction