Learn More
Invitrogen™ SND1 Monoclonal Antibody (2D7)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA515420
Description
MA5-15420 targets SND1/P100 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15420 immunogen is purified recombinant fragment of SND1 (aa361-485) expressed in E. Coli.. MA5-15420 detects SND1/P100 which has a predicted molecular weight of approximately 102kDa.
SND1/P100 (staphylococcal nuclease and tudor domain containing 1), also known as TudorSN, functions in the Pim-1 regulation of Myb activity and acts as a transcriptional activator of EBNA-2. It also interacts with EAV, NSP1, GTF2E1 and GTF2E2, and forms a ternary complex with Stat6 and POLR2A. The staphylococcal nuclease-like (SN)-domains directly interact with amino acids 1099-1758 of CBP. SND1/P100 plays an important role in the assembly of Stat6 transcriptome and stimulates IL-4-dependent transcription by mediating interaction between Stat6 and CBP.
Specifications
SND1 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
Q7KZF4 | |
Snd1 | |
Purified recombinant fragment of SND1 (aa361-485) expressed in E. Coli. | |
RUO | |
27044 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
2D7 | |
Unconjugated | |
Snd1 | |
100 kDa coactivator; 4SNc-Tudor domain protein; 4SNc-Tudor domain protein long form; 4SNc-Tudor domain protein short form; AL033314; EBNA-2 co-activator (100kD); EBNA2 coactivator p100; p100; p100 co-activator; p100 co-activator variant 1; p100 EBNA2 co-activator; p100L; P100-like protein short variant; p100S; similar to Homo sapiens EBNA-2 co-activator (100kDa coactivator); p105 coactivator; SN4TDR; SND p102; SND1; snd1 protein; SND1-BRAF fusion; staphylococcal nuclease and tudor domain containing 1; staphylococcal nuclease domain containing 1; staphylococcal nuclease domain-containing protein 1; TDRD11; testis tissue sperm-binding protein Li 82P; tudor domain-containing protein 11; TudorSN; Tudor-SN; wu:fc27g10 | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.