Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SND1 Mouse anti-Human, Mouse, Clone: 1A6A4, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | SND1 |
---|---|
Clone | 1A6A4 |
Concentration | 1 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6362
![]() ![]() |
Proteintech Group Inc
602651IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
SND1/P100 (staphylococcal nuclease and tudor domain containing 1), also known as TudorSN, it functions in the Pim-1 regulation of Myb activity and acts as a transcriptional activatior of EBNA-2. It also interacts with EAV, NSP1,GTF2E1 and GTF2E2, and forms a ternary complex with Stat6 and POLR2A. The staphylococcal nuclease-like (SN)-domains directly interact with amino acids 1099-1758 of CBP. SND1/P100 plays an important role in the assembly of Stat6 transcriptome and stimulates IL-4-dependent transcription by mediating interaction between Stat6 and CBP.Specifications
SND1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
100 kDa coactivator, EBNA2 coactivator p100, p100, p100 co activator, SND1, TDRD11 | |
SND1 | |
IgG1 | |
Protein G | |
Antibody |
1A6A4 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
Q78PY7, Q7KZF4 | |
27044, 56463 | |
SND1 Fusion Protein Ag1200 | |
Primary | |
-20°C | |
SND1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title