Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SND1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540562
Description
Peptide sequence: LPDYYLVTVM LSGIKCPTFR READGSETPE PFAAEAKFFT ESRLLQRDVQ Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
SND1/P100 (staphylococcal nuclease and tudor domain containing 1), also known as TudorSN, functions in the Pim-1 regulation of Myb activity and acts as a transcriptional activator of EBNA-2. It also interacts with EAV, NSP1, GTF2E1 and GTF2E2, and forms a ternary complex with Stat6 and POLR2A. The staphylococcal nuclease-like (SN)-domains directly interact with amino acids 1099-1758 of CBP. SND1/P100 plays an important role in the assembly of Stat6 transcriptome and stimulates IL-4-dependent transcription by mediating interaction between Stat6 and CBP.
Specifications
SND1 | |
Polyclonal | |
Unconjugated | |
Snd1 | |
100 kDa coactivator; 4SNc-Tudor domain protein; 4SNc-Tudor domain protein long form; 4SNc-Tudor domain protein short form; AL033314; EBNA-2 co-activator (100kD); EBNA2 coactivator p100; p100; p100 co-activator; p100 co-activator variant 1; p100 EBNA2 co-activator; p100L; P100-like protein short variant; p100S; similar to Homo sapiens EBNA-2 co-activator (100kDa coactivator); p105 coactivator; SN4TDR; SND p102; SND1; snd1 protein; SND1-BRAF fusion; staphylococcal nuclease and tudor domain containing 1; staphylococcal nuclease domain containing 1; staphylococcal nuclease domain-containing protein 1; TDRD11; testis tissue sperm-binding protein Li 82P; tudor domain-containing protein 11; TudorSN; Tudor-SN; wu:fc27g10 | |
Rabbit | |
Protein A | |
RUO | |
27044 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q7KZF4 | |
Snd1 | |
Synthetic peptide directed towards the N-terminal of human SND1 (aa 213-262). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction