Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNED1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557648
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557648 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557648 Supplier Invitrogen™ Supplier No. PA557648
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GVRVSIRHPE ALRDQATDVD RSVDRFTFRA LLPGKRYTIQ LTTLSGLRGE EHPTESLATA PTHVWTRPLP PANLTAARVT ATSA Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 70%.

SNED1 (Sushi, Nidogen And EGF Like Domains 1) is a Protein Coding gene. Diseases associated with SNED1 include Cortical Thymoma and Hyperpituitarism. Gene Ontology (GO) annotations related to this gene include calcium ion binding. An important paralog of this gene is NOTCH1. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNED1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SNED1
Gene Accession No. Q8TER0
Gene Alias 6720455I24Rik; 6720455I24Rik homolog; AI197264; AI642697; D430044C15Rik; insulin responsive sequence DNA binding protein-1; insulin-responsive sequence DNA-binding protein 1; IRE-BP1; secreted nidogen domain protein; secreted protein SST3; secreted protein SST-3; Sned1; Snep; SST3; Stromal nidogen extracellular matrix protein; sushi, nidogen and EGF like domains 1; sushi, nidogen and EGF-like domain-containing protein 1; sushi, nidogen and EGF-like domains 1
Gene Symbols SNED1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SNED1. Recombinant protein control fragment (Product #RP-96166).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25992
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.