Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BenchPro™ Sneeze-Guard, Edge Mounted
SDPSCI_ED

Catalog No. S81604
Click to view available options
Height (English):
18 in.
Width (English):
24 in.

Catalog No. S81604

Supplier: BenchPro™ GPE1824/FSE

Get protected while you are working with this BenchPro™ Sneeze Guard Panel. It is easy to setup and easy to clean.

Clean with soap and water. Antibacterial sprays are ok to use as well.

Specifications

Height (English) 18 in.
Width (English) 24 in.
Lens Tint Clear
Product Type Sneeze Guard
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.