Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNF5 Antibody (CL14017) [Alexa Fluor 750], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP343863AF750
Description
SNF5 Monoclonal antibody specifically detects SNF5 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)Specifications
SNF5 | |
Monoclonal | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry (Paraffin) | |
CL14017 | |
Western Blot, Immunohistochemistry-Paraffin | |
BAF47, BAF47hSNF5, BRG1-associated factor 47, hSNFS, Ini1, INI1RTPS1, Integrase interactor 1 protein, malignant rhabdoid tumor suppressor, RDTSNF5 homolog, Sfh1p, SNF5, SNF5L1, Snr1, subfamily b, member 1, sucrose nonfermenting, yeast, homolog-like 1, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily B member 1 | |
This antibody was generated using a recombinant protein sequence of Q12824, with the exact immunogen sequence remaining proprietary. | |
0.1 mL | |
Cell Cycle and Replication | |
6598 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction