Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SNIP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552676
Description
Immunogen sequence: NDLEKSVEKI QRDVSHNHRL VPGPELEEKA LVLKQLGETL TELKAHFPGL QSKMRVVLRV EVEAVKFLKE EPQRLDGLLK RCRGVTDTLA QIRR Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 98%.
Acts as a negative regulator of SRC by activating CSK which inhibits SRC activity and downstream signaling, leading to impaired cell spreading and migration. Regulates dendritic spine morphology. Involved in calcium-dependent exocytosis. May play a role in neurotransmitter release or synapse maintenance.
Specifications
SNIP | |
Polyclonal | |
Unconjugated | |
SRCIN1 | |
KIAA1684; mKIAA1684; p130Cas-associated protein; P140; p140Cap; SNAP-25-interacting protein; SNAP25-interacting protein; SNIP; SRC kinase signaling inhibitor 1; SRCIN1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80725 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9C0H9 | |
SRCIN1 | |
Recombinant protein corresponding to Human SNIP. Recombinant protein control fragment (Product #RP-90631). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction