Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNIP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564135
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564135 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564135 Supplier Invitrogen™ Supplier No. PA564135
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EGVWPPPNNL LSQSPKKVTA ETDFNKSVDF EMPPPSPPLN LHELSGPAEG ASLTPKGGNP TKGLDTPGKR SVDKAVSVEA AERDWE Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 85%.

Acts as a negative regulator of SRC by activating CSK which inhibits SRC activity and downstream signaling, leading to impaired cell spreading and migration. Regulates dendritic spine morphology. Involved in calcium-dependent exocytosis. May play a role in neurotransmitter release or synapse maintenance.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNIP
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SRCIN1
Gene Accession No. Q9C0H9
Gene Alias KIAA1684; mKIAA1684; p130Cas-associated protein; P140; p140Cap; SNAP-25-interacting protein; SNAP25-interacting protein; SNIP; SRC kinase signaling inhibitor 1; SRCIN1
Gene Symbols SRCIN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SNIP. Recombinant protein control fragment (Product #RP-103528).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80725
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.