Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SNIP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5102007
Description
Antibody detects endogenous levels of total SNIP.
Acts as a negative regulator of SRC by activating CSK which inhibits SRC activity and downstream signaling, leading to impaired cell spreading and migration. Regulates dendritic spine morphology. Involved in calcium-dependent exocytosis. May play a role in neurotransmitter release or synapse maintenance.
Specifications
SNIP | |
Polyclonal | |
Unconjugated | |
SRCIN1 | |
KIAA1684; mKIAA1684; p130Cas-associated protein; P140; p140Cap; SNAP-25-interacting protein; SNAP25-interacting protein; SNIP; SRC kinase signaling inhibitor 1; SRCIN1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
56013, 56029, 80725 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9C0H9, Q9QWI6, Q9QXY2 | |
SRCIN1 | |
A synthesized peptide derived from human SRCIN1(Accession Q9C0H9), corresponding to amino acid residues K943-P993. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction