Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SNIP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA529412
Description
Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Mouse (86%), Rat (87%), Rhesus Monkey (97%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Smad-binding peptide aptamers can be developed to selectively inhibit TGF-beta-induced gene expression.
Specifications
SNIP1 | |
Polyclonal | |
Unconjugated | |
SNIP1 | |
2410133M08Rik; dJ423B22.2; FHA domain-containing protein SNIP1; PMRED; RP3-423B22.3; Smad nuclear interacting protein; smad nuclear interacting protein 1; Smad nuclear interacting protein 1 Snip1; smad nuclear-interacting protein 1; Snip1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
313588, 76793, 79753 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q5M9G6, Q8BIZ6, Q8TAD8 | |
SNIP1 | |
Recombinant fragment corresponding to a region within amino acids 41 and 363 of Human SNIP1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction