Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SNIP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116692
Description
Positive Samples: BxPC-3 Immunogen sequence: SESQELVPRP GGNNKEKEVP AKEKPSFELS GALLEDTNTF RGVVIKYSEP PEARIPKKRW RLYPFKNDEV LPVMYIHRQS AYLLGRHRRI ADIPIDHPSC S.
Smad-binding peptide aptamers can be developed to selectively inhibit TGF-beta-induced gene expression.
Specifications
SNIP1 | |
Polyclonal | |
Unconjugated | |
SNIP1 | |
2410133M08Rik; dJ423B22.2; FHA domain-containing protein SNIP1; PMRED; RP3-423B22.3; Smad nuclear interacting protein; smad nuclear interacting protein 1; Smad nuclear interacting protein 1 Snip1; smad nuclear-interacting protein 1; Snip1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
313588, 79753 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
4.47 mg/mL | |
PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
Q5M9G6, Q8TAD8 | |
SNIP1 | |
A synthetic peptide corresponding to a sequence within amino acids 200-300 of human SNIP1 (NP_0789762). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction