Learn More
SNM1A Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303746A

Description
The recommended shelf life for this product is 1 year from date of receipt.
May be required for DNA interstrand cross-link repair. Also required for checkpoint mediated cell cycle arrest in early prophase in response to mitotic spindle poisons.Specifications
SNM1A | |
Polyclonal | |
Unconjugated | |
DCLRE1A | |
PSO2, SNM1, SNM1A, DNA cross-link repair 1A (PSO2 homolog, S. cerevisiae), DNA cross-link repair 1A protein, DNA-crosslink repair gene SNM1, PSO2 homolog, SNM1 homolog A, hSNM1A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9937 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q6PJP8 | |
DCLRE1A | |
Between 1 and 50 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.