Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SnoN Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP224718SS
Description
SnoN Polyclonal specifically detects SnoN in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
SnoN | |
Polyclonal | |
Unconjugated | |
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
SKIL | |
A portion of amino acids 600-684 of human snoN protein were used as the immunogen. | |
0.025 mg | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
0.5 mg/mL | |
Western Blot 1-3 ug/ml | |
SKI-like oncogene, Ski-related oncogene, ski-related oncogene snoN, Ski-related protein, SnoA, SnoI, SnoN, SNOski-like protein | |
Rabbit | |
Protein G purified | |
RUO | |
6498 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction