Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SnoN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598150
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598150 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598150 Supplier Invitrogen™ Supplier No. PA598150
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SnoN Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

TGF-beta is a ubiquitously expressed cytokine that signals through the Smad protein family to regulate numerous cellular processes such as embryonic development and tumorigenesis. This signaling is negatively regulated by Ski, the mammalian homolog of the transforming protein of an avian retrovirus that induces oncogenic transformation of chicken embryo cells, and the related protein SnoN. Like Ski, SnoN functions by binding to the Smad proteins and preventing their phosphorylation, thereby inhibiting their ability to bind DNA and activate the transcription of downstream genes. SnoN is located primarily in the nucleus in cancer tissues or cells, but in the cytoplasm in normal tissues or primary epithelial cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SnoN
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SKIL
Gene Accession No. P12757, Q60665
Gene Alias EGK_11909; LOW QUALITY PROTEIN: ski-like protein; SKI like proto-oncogene; ski/sno related; SKIL; SKI-like; SKI-like oncogene; ski-like protein; ski-like protein-like; SKI-like proto-oncogene; Skir; ski-related oncogene; ski-related oncogene snoN; Ski-related protein; SNO; SnoA; SnoI; SnoN; v-ski sarcoma viral oncogene homolog
Gene Symbols SKIL
Host Species Rabbit
Immunogen Recombinant Human Ski-like protein (359-579AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114208, 20482, 6498
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.