Learn More
Copan Diagnostics™ SnotBuster™ Sputum Liquefying System
SDP
Catalog No. p-8750019 Shop All Copan Diagnostics Inc. Products
Click to view available options
Includes:
SLSolution in PET Tube
SLSolution, Tube, Transfer Device
Transfer Device
This item is not returnable. View return policy
Catalog No. 23-600-526
Supplier: Copan Diagnostics™ 0U019N
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Save cost and avoid waste of making your own reagents for liquifying sputum with this single use product
  • SnotBuster™ (a.k.a. SLsolution) is a ready to use mucolytic agent for sputum samples with the principle active ingredient being Dithiotreitol (DTT)
  • SnotBuster™ has been tested and validated for liquefying sputum samples prior to culturing for the isolation of bacteria and fungi without affecting the morphology, growth or microscopic staining and appearance of pathogens
  • Liquid based system is easily handled by automated specimen processors and automatic pipetting systems, minimizing manual handling
  • Easier, more consistent and reproducible specimen planting and streaking
  • Simplify handling of challenging sputum specimens using the efficient Sputum Dipper™ tool, handles all types of sputum viscosity from liquid to very tenacious sputum samples
  • The ingenious drill-shaped design of the Sputum Dipper™ allows laboratory technologists to easily manipulate and transfer a representative sample of sputum from the sample container to SnotBuster™
Specifications
Sterility Sterile
For Use With (Application) Liquefying Sputum Specimens
Includes SLSolution in PET Tube
Quantity 50/Pk, 300/Cs.
Volume (Metric) 1 mL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.