Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNRNP27 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548302
Description
May play a role in mRNA splicing.
Specifications
SNRNP27 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q8WVK2, Q8K194 | |
SNRNP27 | |
KLH conjugated synthetic peptide between 69-98 amino acids from the Central region of human SNR27. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
SNRNP27 | |
U4/U6U5 small nuclear ribonucleoprotein 27 kDa protein, U4/U6U5 snRNP 27 kDa protein, U4/U6U5-27K, Nucleic acid-binding protein RY-1, U4/U6U5 tri-snRNP-associated 27 kDa protein, 27K, U4/U6U5 tri-snRNP-associated protein 3, SNRNP27 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11017, 66618 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction