Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNRK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544539
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544539 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544539 Supplier Thermo Scientific Supplier No. PA544539
Add to Cart
Add to Cart

SNRK Polyclonal Antibody

SNRK Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen SNRK
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SNRK
Gene Accession No. Q9NRH2
Gene Alias 2010012F07Rik; AI448042; AW547029; E030034B15; HSNFRK; KIAA0096; mKIAA0096; R74830; RGD:69653}; SNF related kinase; SNF-1 related kinase; SNF1-related kinase; SNF-related serine/threonine-protein kinase; SNF-related serine/threonine-protein kinase; LOW QUALITY PROTEIN: SNF-related serine/threonine-protein kinase; SNFRK; SNRK; snrk {ECO:0000312
Gene Symbols SNRK
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human SNRK.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54861
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.